Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
qtc
Entrez Gene ID
33739
Gene Name
quick-to-court
Gene Aliases
CG14039, Dmel\CG14039, Dmel_CG14039
Chromosome Location
Chr.2L: 5060815 - 5070750 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0028572
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014467.2 NP_001014467.1 2 - 3 645 167
NM_001014469.2 NP_001014469.1 2 - 3 645 167
NM_001273155.1 NP_001260084.1 1 - 2 238 167
NM_078756.4 NP_523480.2 2 - 3 645 167
NM_164627.2 NP_723059.2 1 - 2 238 167
GenBank mRNA AY058777.1 - 2 - 3 628 167

View Full Product Details