Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GluRIIA
- Entrez Gene ID
-
33788
- Gene Name
- Glutamate receptor IIA
- Gene Aliases
-
CG6992, D-GluRIIA, DGluR-II, DGluR-IIA, DGluR2a, DGluRIIA, DGluRIIa, DgluRII, DmelGluRIIA, Dmel\CG6992, Dmel_CG6992, Glu-RII, Glu-RIIA, GluR, GluR-IIA, GluR2, GluR2a, GluRII, GluRII-A, GluRIIa, GluR[[IIa]], GlurIIA, dGluRII, dGluRIIA, dglur-IIA, dglurIIA, gluIIA, gluRIIA, glurIIA, gluriia
- Chromosome Location
-
Chr.2L: 5555072 - 5558994 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0004620
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078760.3 |
NP_523484.2 |
4 - 5 |
756 |
73 |
GenBank mRNA |
BT010006.1 |
- |
4 - 5 |
754 |
73 |
View Full Product Details