Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- GluRIIB
- Entrez Gene ID
-
33789
- Gene Name
- Glutamate receptor IIB
- Gene Aliases
-
CG7234, DGluIIB, DGluR-IIB, DGluRIIB, DmelGluRIIB, Dmel\CG7234, Dmel_CG7234, Glu-RIIB, GluRIIb, dGluRIIB, dglurIIB, glurIIB
- Chromosome Location
-
Chr.2L: 5559342 - 5563502 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0020429
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078761.3 |
NP_523485.3 |
4 - 5 |
609 |
68 |
View Full Product Details