Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cup
Entrez Gene ID
33934
Gene Name
CG11181-PB
Gene Aliases
CG11181, CUP, Cup, DmCup, Dmel\CG11181, Dmel_CG11181, fs(1)cup, fs(2)cup, i239
Chromosome Location
Chr.2L: 6663664 - 6674780 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0000392
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001298758.1 NP_001285687.1 1 - 2 923 68
NM_078769.3 NP_523493.3 1 - 2 619 68

Gene Symbol
CG34310
Entrez Gene ID
5740536
Gene Name
CG34310-PA
Gene Aliases
Dmel\CG34310, Dmel_CG34310
Chromosome Location
Chr.2L: 6663664 - 6674780 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0085339
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103634.2 NP_001097104.2 1 - 2 619 68
NM_001298757.1 NP_001285686.1 1 - 2 923 68

View Full Product Details