Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arpc2
Entrez Gene ID
35311
Gene Name
Actin-related protein 2/3 complex, subunit 2
Gene Aliases
38C.47, ARC-P34, ARC-p34, ARPC2, ARPC2/p34, Arc-p34, Arc-p34/Arpc2, ArcP34, CG10954, Dmel\CG10954, Dmel_CG10954, anon-WO0118547.154, p34
Chromosome Location
Chr.2L: 20417057 - 20423027 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0032859
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001299173.1 NP_001286102.1 7 - 8 1409 154
NM_136189.4 NP_610033.1 7 - 8 1376 154
GenBank mRNA AY060391.1 - 7 - 8 1352 154

View Full Product Details