Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Oseg5
- Entrez Gene ID
-
35349
- Gene Name
- Outer segment 5
- Gene Aliases
-
CG9333, CHE-2, Che-2, Dmel\CG9333, Dmel_CG9333, IFT80, OSEG, OSEG5, oseg5
- Chromosome Location
-
Chr.2L: 20832849 - 20835840 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0032891
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136220.3 |
NP_610064.2 |
1 - 2 |
366 |
80 |
View Full Product Details