Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
step
Entrez Gene ID
35425
Gene Name
steppke
Gene Aliases
CG11628, CG11633, CYH1, Dmel\CG11628, Dmel_CG11628, GPH, GRP1, GRP1/cytohesin 1, Grp1, Step, cytohesin/GRP1, l(2)SH0323, l(2)SH2 0323, l(2)k08110, stepk
Chromosome Location
Chr.2L: 21740041 - 21757466 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0086779
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042910.3 NP_001036375.2 6 - 7 1758 73
NM_001169570.2 NP_001163041.1 5 - 6 2019 73
NM_001273749.1 NP_001260678.1 7 - 8 2487 73
NM_136276.4 NP_610120.1 5 - 6 710 73
GenBank mRNA AY071322.1 - 5 - 6 644 73
BT072836.1 - 5 - 6 662 73
BT124891.1 - 7 - 8 2386 73

View Full Product Details