Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Src42A
- Entrez Gene ID
-
35524
- Gene Name
- Src oncogene at 42A
- Gene Aliases
-
CG14471, CG44128, CG7873, DSrc42A, Dm SRC41, DmHD-29, Dmel\CG44128, Dmel_CG44128, Dsrc41, Dsrc42A, Dtk5, HD-29, SK2-4, SRC 42A, SRC42A, Src, Src41, Src42, Src42a, Su(D-raf)1, Su(Raf)1, Su(phl)1, Su1, Tk5, dSrc, dSrc42A, dtk-5, dtk5, l(2)k10108, src, src42, src42A, src42a
- Chromosome Location
-
Chr.2R: 5981844 - 6012795 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0264959
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136347.2 |
NP_610191.2 |
2 - 2 |
4000 |
59 |
View Full Product Details