Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ptr
Entrez Gene ID
35546
Gene Name
Patched-related
Gene Aliases
CG11212, Dmel\CG11212, Dmel_CG11212, Dmptr, Ptr_Dm, ptc-related-gene, ptr
Chromosome Location
Chr.2R: 6181748 - 6196320 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0262867
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259216.2 NP_001246145.1 7 - 8 1631 86
NM_001259217.2 NP_001246146.1 7 - 8 1687 86
NM_136365.3 NP_610209.2 7 - 8 1376 86
GenBank mRNA BT003500.1 - 7 - 8 1374 86

View Full Product Details