Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tdc1
- Entrez Gene ID
-
35573
- Gene Name
- Tyrosine decarboxylase 1
- Gene Aliases
-
CG30445, CG3686, Dmel\CG30445, Dmel_CG30445, TDC, dTdc1, tdc, tdc1
- Chromosome Location
-
Chr.2R: 6677001 - 6680391 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0259977
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136382.3 |
NP_610226.2 |
2 - 3 |
1449 |
64 |
GenBank mRNA |
BT044119.1 |
- |
2 - 3 |
1444 |
64 |
View Full Product Details