Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spn43Ad
Entrez Gene ID
35639
Gene Name
Serpin 43Ad
Gene Aliases
CG1859, Dmel\CG1859, Dmel_CG1859
Chromosome Location
Chr.2R: 7153738 - 7155424 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0044011
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136417.3 NP_610261.1 2 - 3 1183 58
GenBank mRNA AY071238.1 - 2 - 3 1181 58

View Full Product Details