Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hrg
Entrez Gene ID
49636
Gene Name
hiiragi
Gene Aliases
CG9854, Dmel\CG9854, Dmel_CG9854, PAP, Pap, dHrg, l(2)07619, l(2)k07619
Chromosome Location
Chr.2R: 19663919 - 19669832 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015949
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043097.3 NP_001036562.1 5 - 6 2248 85
NM_001274168.2 NP_001261097.1 5 - 6 2377 85
NM_080529.4 NP_536790.1 5 - 6 2248 85
NM_166372.3 NP_725919.1 5 - 6 1941 85
GenBank mRNA AB018431.1 - 5 - 6 2237 85
BT001541.1 - 5 - 6 1934 85
BT050444.1 - 5 - 6 2245 85

View Full Product Details