Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hrg
Entrez Gene ID
49636
Gene Name
hiiragi
Gene Aliases
CG9854, Dmel\CG9854, Dmel_CG9854, PAP, Pap, dHrg, l(2)07619, l(2)k07619
Chromosome Location
Chr.2R: 19663919 - 19669832 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015949
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043097.3 NP_001036562.1 6 - 7 2525 61
NM_001274168.2 NP_001261097.1 6 - 7 2654 61
NM_080529.4 NP_536790.1 6 - 7 2525 61
NM_166372.3 NP_725919.1 6 - 7 2218 61
GenBank mRNA AB018431.1 - 6 - 7 2514 61
AF231704.1 - 7 - 8 2484 61
BT001541.1 - 6 - 7 2211 61
BT050444.1 - 6 - 7 2522 61

View Full Product Details