Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hrg
Entrez Gene ID
49636
Gene Name
hiiragi
Gene Aliases
CG9854, Dmel\CG9854, Dmel_CG9854, PAP, Pap, dHrg, l(2)07619, l(2)k07619
Chromosome Location
Chr.2R: 19663919 - 19669832 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0015949
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274168.2 NP_001261097.1 7 - 8 3351 127
NM_080529.4 NP_536790.1 7 - 8 3222 127
NM_166372.3 NP_725919.1 7 - 8 2915 127
GenBank mRNA AF231704.1 - 8 - 9 3181 127
BT001541.1 - 7 - 8 2908 127
BT050444.1 - 7 - 8 3219 127

View Full Product Details