Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Rx
- Entrez Gene ID
-
37367
- Gene Name
- Retinal Homeobox
- Gene Aliases
-
CG10052, DRx, Dmel\CG10052, Dmel_CG10052, Drx, E97, PPH17, W60, bk50, drx, rx, wom
- Chromosome Location
-
Chr.2R: 20916753 - 20937030 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0020617
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166413.3 |
NP_726006.3 |
2 - 3 |
817 |
56 |
View Full Product Details