Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Xpd
- Entrez Gene ID
-
37414
- Gene Name
- Xeroderma pigmentosum D
- Gene Aliases
-
CG9433, DhR3, DhXPD, DmXPD, Dmel\CG9433, Dmel_CG9433, ERCC2, XPD, XPD/ERCC2, l(2)SH2 2137, l(2)SH2137, xpd
- Chromosome Location
-
Chr.2R: 21171001 - 21174504 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0261850
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166429.3 |
NP_726036.2 |
5 - 6 |
1263 |
76 |
View Full Product Details