Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Tim10
- Entrez Gene ID
-
37478
- Gene Name
- Translocase of inner membrane 10
- Gene Aliases
-
CG9878, Dmel\CG9878, Dmel_CG9878, Dromel_CG9878_FBtr0071676_tim10_mORF, bs31d12.y1, tim10
- Chromosome Location
-
Chr.2R: 21690215 - 21691139 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0027360
- Gene Symbol
- CG42497
- Entrez Gene ID
-
8674071
- Gene Name
- CG42497-PA
- Gene Aliases
-
Dmel\CG42497, Dmel_CG42497, Dromel_CG9878_FBtr0071676_tim10_uORF
- Chromosome Location
-
Chr.2R: 21690215 - 21691139 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0260223
View Full Product Details