Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
px
Entrez Gene ID
37573
Gene Name
plexus
Gene Aliases
CG4444, Dmel\CG4444, Dmel_CG4444, l(2)k08134, l(2)k08316
Chromosome Location
Chr.2R: 22494297 - 22562065 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0003175
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274207.1 NP_001261136.1 3 - 4 2143 54
NM_001382123.1 NP_001369111.1 3 - 4 2123 54
NM_001382124.1 NP_001369112.1 3 - 4 2123 54
NM_166535.2 NP_726208.1 3 - 4 2123 54

View Full Product Details