Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GABA-B-R1
Entrez Gene ID
34878
Gene Name
metabotropic GABA-B receptor subtype 1
Gene Aliases
BG:DS00929.6, CG15274, D-GABA[[B]]R1, D-Gaba1, Dmel\CG15274, Dmel_CG15274, GABA(B)R1, GABA-BR1, GABA-[[B]]-R1, GABA[[B]], GABA[[B]]-R, GABA[[B]]-R1, GABA[[B]]R, GABA[[B]]R1, GB1, anon-WO0170980.175, anon-WO0170980.176
Chromosome Location
Chr.2L: 15016331 - 15034704 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260446
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001259104.2 NP_001246033.1 8 - 9 2399 97
NM_078845.5 NP_523569.3 8 - 9 2399 97
GenBank mRNA BT031099.1 - 8 - 9 1595 97

View Full Product Details