The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001382179.1 | NP_001369131.1 | 6 - 7 | 2924 | 66 |
NM_078867.4 | NP_523591.2 | 6 - 7 | 2924 | 66 |