Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- bsf
- Entrez Gene ID
-
35100
- Gene Name
- bicoid stability factor
- Gene Aliases
-
BSF, CG10302, DmLRPPRC1, DmLrpprc1, Dmel\CG10302, Dmel_CG10302, dmlrpprc1, l(2)SH1181, l(2)SH2 1181
- Chromosome Location
-
Chr.2L: 18449517 - 18454587 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0284256
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078872.5 |
NP_523596.2 |
1 - 2 |
245 |
84 |
GenBank mRNA |
BT150103.1 |
- |
1 - 2 |
228 |
84 |
View Full Product Details