Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Top3alpha
- Entrez Gene ID
-
35236
- Gene Name
- Topoisomerase 3alpha
- Gene Aliases
-
CG10123, DmTop3alpha, Dmel\CG10123, Dmel_CG10123, Top3a, Topo IIIalpha, dTopoIIIa, top3alpha, topo IIIalpha
- Chromosome Location
-
Chr.2L: 19498168 - 19502268 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0040268
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078878.3 |
NP_523602.2 |
2 - 3 |
867 |
70 |
GenBank mRNA |
BT015282.1 |
- |
2 - 3 |
840 |
70 |
View Full Product Details