Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- phr
- Entrez Gene ID
-
35735
- Gene Name
- photorepair
- Gene Aliases
-
C2PHR, CG11205, CG11205-PA, Dm CPD, Dmel\CG11205, Dmel_CG11205, Phl, anon-OV1, anonOV1
- Chromosome Location
-
Chr.2R: 7825289 - 7827319 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0003082
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_078929.4 |
NP_523653.2 |
1 - 2 |
260 |
65 |
GenBank mRNA |
AY084133.1 |
- |
1 - 2 |
52 |
65 |
View Full Product Details