Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cul4
Entrez Gene ID
35780
Gene Name
Cullin 4
Gene Aliases
CG8711, CUL4, Cul-4, Cullin4, DMcul-4, Dmel\CG8711, Dmel_CG8711, cul-4, cul4, dCul4
Chromosome Location
Chr.2R: 8102260 - 8106318 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033260
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169613.1 NP_001163084.1 7 - 8 1941 129
NM_136508.4 NP_610352.2 7 - 8 1945 129
GenBank mRNA BT021374.1 - 7 - 8 1899 129

View Full Product Details