Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pbp49
Entrez Gene ID
35787
Gene Name
PSEA-binding protein 49kD
Gene Aliases
CG11508, CG42515, DMPBP49, DmPBP, DmPBP49, DmSNAP50, DmSNAPc, Dmel\CG42515, Dmel_CG42515, Dromel_CG11508_FBtr0088817_mORF
Chromosome Location
Chr.2R: 8129354 - 8131323 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260398
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144144.2 NP_001137616.2 2 - 2 1773 92
NM_136513.4 NP_610357.1 1 - 1 1830 92
NM_165588.4 NP_724647.1 2 - 2 1747 92
GenBank mRNA AY119595.1 - 1 - 1 1813 92

Gene Symbol
CG42516
Entrez Gene ID
8674068
Gene Name
CG42516-PA
Gene Aliases
Dmel\CG42516, Dmel_CG42516, Dromel_CG11508_FBtr0088817_uORF
Chromosome Location
Chr.2R: 8129354 - 8131323 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0260390
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169615.2 NP_001163086.1 2 - 2 1747 92
NM_001169616.2 NP_001163087.1 2 - 2 1747 92
NM_001299256.1 NP_001286185.1 2 - 2 1773 92

View Full Product Details