Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
pdm3
Entrez Gene ID
35813
Gene Name
pou domain motif 3
Gene Aliases
BcDNA:AT16994, CG11641, CG14755, CG42698, Dmel\CG42698, Dmel_CG42698, Pdm3
Chromosome Location
Chr.2R: 8327490 - 8396280 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0261588
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144145.4 NP_001137617.2 4 - 5 2333 62
NM_001201956.2 NP_001188885.2 4 - 5 2333 62
NM_001299264.1 NP_001286193.1 4 - 5 2333 62
NM_136533.2 NP_610377.1 3 - 4 1246 62
GenBank mRNA AY122074.1 - 3 - 4 1246 62

View Full Product Details