Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Cyp4ad1
- Entrez Gene ID
-
35821
- Gene Name
- CG2110-PA
- Gene Aliases
-
4ad1, BEST:LD02646, CG2110, Cyp4AD1, Dmel\CG2110, Dmel_CG2110
- Chromosome Location
-
Chr.2R: 8439395 - 8442806 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033292
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_136536.3 |
NP_610380.1 |
1 - 2 |
220 |
64 |
GenBank mRNA |
AY061058.1 |
- |
1 - 2 |
193 |
64 |
View Full Product Details