Assay Design
The assay primers and probes lie within a single exon. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- PGRP-SC1b
- Entrez Gene ID
-
35861
- Gene Name
- Peptidoglycan recognition protein SC1b
- Gene Aliases
-
CG8577, CT8705, Dmel\CG8577, Dmel_CG8577, PGRP SC, PGRP-SC, PGRP-SC1, PGRP-SC1B, SC1B, pgrp-sc, pgrp-sc1
- Chromosome Location
-
Chr.2R: 8713446 - 8715094 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0033327
- Gene Symbol
- PGRP-SC1a
- Entrez Gene ID
-
35859
- Gene Name
- Peptidoglycan recognition protein SC1a
- Gene Aliases
-
CG14746, Dmel\CG14746, Dmel_CG14746, PGRP SC, PGRP-SC, PGRP-SC1, PGRP-SC1A, pgrp-sc, pgrp-sc1, picky
- Chromosome Location
-
Chr.2R: 8709733 - 8710320 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0043576
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AF207542.1 |
- |
1 - 1 |
570 |
70 |
BT044352.1 |
- |
1 - 1 |
569 |
70 |
View Full Product Details