Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cyp4p2
Entrez Gene ID
35946
Gene Name
CG1944-PA
Gene Aliases
4p2, CG1944, CYP4P2, Dmel\CG1944, Dmel_CG1944
Chromosome Location
Chr.2R: 9237512 - 9239831 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033395
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_136628.3 NP_610472.1 2 - 3 444 120
GenBank mRNA AY051564.1 - 2 - 3 426 120

View Full Product Details