Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pde8
Entrez Gene ID
37741
Gene Name
Phosphodiesterase 8
Gene Aliases
CG42241, CG45019, CG5411, Dmel\CG45019, Dmel_CG45019, PDE8, pde8
Chromosome Location
Chr.2R: 23657567 - 23680844 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0266377
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169803.2 NP_001163274.1 9 - 10 1510 85
NM_001169804.2 NP_001163275.2 5 - 6 1670 85
NM_001169805.2 NP_001163276.1 8 - 9 1337 85
NM_001169807.2 NP_001163278.2 8 - 9 1623 85
NM_137970.5 NP_611814.3 9 - 10 1368 85
NM_166616.3 NP_726350.1 8 - 9 1623 85
NM_166617.2 NP_726351.1 6 - 7 757 85
NM_166618.3 NP_726352.1 7 - 8 1106 85
GenBank mRNA BT011336.1 - 8 - 9 1587 85
BT021326.1 - 9 - 10 1364 85
BT133111.1 - 7 - 8 1106 85

View Full Product Details