Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Pde8
- Entrez Gene ID
-
37741
- Gene Name
- Phosphodiesterase 8
- Gene Aliases
-
CG42241, CG45019, CG5411, Dmel\CG45019, Dmel_CG45019, PDE8, pde8
- Chromosome Location
-
Chr.2R: 23657567 - 23680844 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0266377
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_166618.3 |
NP_726352.1 |
2 - 3 |
610 |
81 |
GenBank mRNA |
BT133111.1 |
- |
2 - 3 |
610 |
81 |
View Full Product Details