Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gol
Entrez Gene ID
38006
Gene Name
goliath
Gene Aliases
CG2679, Dmel\CG2679, Dmel_CG2679, Gol, anon-G1, dgol, g1, meso
Chromosome Location
Chr.2R: 25072627 - 25084347 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0004919
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169829.3 NP_001163300.1 3 - 4 1252 115
NM_001259588.2 NP_001246517.1 3 - 4 1252 115
NM_079140.5 NP_523864.3 3 - 4 1252 115
NM_166705.3 NP_726508.1 3 - 4 1252 115
GenBank mRNA AY069169.1 - 3 - 4 1225 115

View Full Product Details