Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Eph
Entrez Gene ID
43803
Gene Name
Eph receptor tyrosine kinase
Gene Aliases
CG1511, CT3831, DEK, Dek, Dek7, Deph, Dmel\CG1511, Dmel_CG1511, Ek7, RPTK Dek7, dek, eph, lincRNA.923
Chromosome Location
Chr.4: 610684 - 622470 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0025936
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169353.2 NP_001162824.1 4 - 5 584 94
NM_079886.5 NP_524625.3 3 - 4 658 94
NM_166774.3 NP_726588.2 3 - 4 658 94
NM_166775.4 NP_726589.2 3 - 4 658 94
NM_166776.4 NP_726590.2 3 - 4 658 94
NM_166777.4 NP_726591.2 4 - 5 584 94

View Full Product Details