Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
gw
Entrez Gene ID
43808
Gene Name
gawky
Gene Aliases
CG11484, CG31992, CG9905, Dmel\CG31992, Dmel_CG31992, GQ182, GW, GW182, Gw, Gw182, anon-EST:Posey205, dGW182, gw182
Chromosome Location
Chr.4: 649041 - 660608 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0051992
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014691.3 NP_001014691.2 6 - 7 3110 82
NM_166780.2 NP_726596.1 6 - 7 3354 82
NM_166781.2 NP_726597.1 6 - 7 3110 82
NM_166784.2 NP_726600.1 6 - 7 2993 82
NM_166785.2 NP_726601.1 6 - 7 3036 82
GenBank mRNA AY075429.1 - 6 - 7 3308 82

View Full Product Details