Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cals
Entrez Gene ID
43824
Gene Name
Calsyntenin-1
Gene Aliases
CALS, CG11059, CT30877, Cad102F, DCad102F, Dmel\CG11059, Dmel_CG11059, cals
Chromosome Location
Chr.4: 1112874 - 1123357 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0039928
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272157.1 NP_001259086.1 4 - 5 947 82
NM_001297793.1 NP_001284722.1 4 - 5 950 82
NM_079893.3 NP_524632.1 4 - 5 947 82
NM_166823.2 NP_726646.1 4 - 5 950 82
GenBank mRNA AJ289018.1 - 4 - 5 913 82
AY121644.1 - 4 - 5 915 82

View Full Product Details