Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP-alpha
Entrez Gene ID
31322
Gene Name
Dpr-interacting protein alpha
Gene Aliases
32791, CG13019, CG13020, CG32791, Dmel\CG32791, Dmel_CG32791
Chromosome Location
Chr.X: 3507245 - 3571649 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0052791
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001272288.2 NP_001259217.1 9 - 10 2052 71
NM_001272289.1 NP_001259218.1 7 - 8 1088 71
NM_166976.3 NP_726871.1 7 - 8 1088 71
GenBank mRNA BT003528.1 - 7 - 8 1085 71

View Full Product Details