Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fd3F
Entrez Gene ID
31336
Gene Name
forkhead domain 3F
Gene Aliases
CG12632, CG15238, CG18771, CG32779, CG44123, Dmel\CG44123, Dmel_CG44123, Fd3F, fd3f
Chromosome Location
Chr.X: 3802316 - 3811040 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264954
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001370027.1 NP_001356931.1 1 - 1 1078 102
NM_166991.3 NP_726889.3 1 - 1 1078 102

View Full Product Details