Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GLaz
Entrez Gene ID
36447
Gene Name
Glial Lazarillo
Gene Aliases
CG4604, DGLaz, Dmel\CG4604, Dmel_CG4604, dGLaz, glaz
Chromosome Location
Chr.2R: 13163959 - 13168032 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0033799
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274007.2 NP_001260936.1 3 - 4 496 100
NM_001274008.2 NP_001260937.1 3 - 4 615 100
NM_001274009.2 NP_001260938.1 3 - 4 518 100
NM_001274010.2 NP_001260939.1 3 - 4 579 100
NM_001274011.2 NP_001260940.1 3 - 4 501 100
NM_079003.5 NP_523727.2 3 - 4 513 100

View Full Product Details