Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Or59b
- Entrez Gene ID
-
37715
- Gene Name
- Odorant receptor 59b
- Gene Aliases
-
59E.1, 59b, CG3569, DOR119, DmOR59B, DmOr59b, Dmel\CG3569, Dmel_CG3569, OR59b, Or59B
- Chromosome Location
-
Chr.2R: 23470165 - 23471775 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0034865
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079098.2 |
NP_523822.1 |
2 - 3 |
1197 |
88 |
View Full Product Details