Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- mAChR-A
- Entrez Gene ID
-
37892
- Gene Name
- muscarinic Acetylcholine Receptor, A-type
- Gene Aliases
-
Acr60C, AcrC, CG4356, DM1, Dm-mAChR-A, Dm1, Dmel\CG4356, Dmel_CG4356, mAChR, mAChR A, mAChR-1, mAcR, mAcR-60C, mAcR60C, mAchR
- Chromosome Location
-
Chr.2R: 24378630 - 24390294 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0000037
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079120.3 |
NP_523844.2 |
4 - 5 |
2196 |
78 |
View Full Product Details