Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
klar
Entrez Gene ID
38067
Gene Name
klarsicht
Gene Aliases
CG17046, Dmel\CG17046, Dmel_CG17046, Klar, kls, marb
Chromosome Location
Chr.3L: 434083 - 540614 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0001316
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001043111.3 NP_001036576.1 5 - 6 2043 73
NM_001103993.2 NP_001097463.1 5 - 6 2043 73
NM_001274282.2 NP_001261211.1 4 - 5 1949 73
NM_079149.5 NP_523873.2 5 - 6 2043 73
GenBank mRNA BT010042.1 - 5 - 6 1633 73

View Full Product Details