Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Glut1
Entrez Gene ID
38109
Gene Name
Glucose transporter 1
Gene Aliases
0074/12, CG1086, CG13908, CG43946, DmGLUT1, DmGluT1, Dmel\CG43946, Dmel_CG43946, Dmglut1, GLU, GLUT1, Glt1, Glu T1, Glut-1, glut1, l(3)61Ea, l(3)L1H, l(3)S007412
Chromosome Location
Chr.3L: 914074 - 993788 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0264574
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001103997.3 NP_001097467.1 5 - 6 1256 108
NM_001103998.3 NP_001097468.1 5 - 6 1256 108
NM_001103999.4 NP_001097469.2 7 - 8 1560 108
NM_001104000.2 NP_001097470.2 5 - 6 1256 108
NM_001274301.1 NP_001261230.1 5 - 6 1256 108
NM_001274302.1 NP_001261231.1 5 - 6 1256 108
NM_001274303.1 NP_001261232.1 8 - 9 1616 108
NM_001274304.1 NP_001261233.1 13 - 14 2347 108
NM_001274305.1 NP_001261234.1 13 - 14 2928 108
NM_001274307.1 NP_001261236.1 10 - 11 2002 108
NM_001274308.1 NP_001261237.1 10 - 11 2002 108
NM_001274309.1 NP_001261238.1 10 - 11 2002 108
NM_001299966.1 NP_001286895.1 5 - 6 1256 108
NM_001299967.1 NP_001286896.1 5 - 6 1256 108
NM_001316474.1 NP_001303403.1 5 - 6 1256 108
NM_079154.3 NP_523878.1 5 - 6 1256 108
NM_138229.2 NP_612073.2 5 - 6 1256 108
NM_167845.3 NP_728557.1 5 - 6 1128 108
NM_167846.3 NP_728558.5 12 - 13 2194 108
GenBank mRNA AF064703.2 - 5 - 6 1254 108
BT011383.1 - 5 - 6 1224 108

View Full Product Details