Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
yellow-g
Entrez Gene ID
38294
Gene Name
CG5717-PA
Gene Aliases
CG5717, Dmel\CG5717, Dmel_CG5717, g, yell-g
Chromosome Location
Chr.3L: 2269881 - 2272769 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0041709
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079164.3 NP_523888.1 2 - 3 723 83
GenBank mRNA BT150079.1 - 2 - 3 723 83

View Full Product Details