Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Or63a
Entrez Gene ID
38354
Gene Name
Odorant receptor 63a
Gene Aliases
63B.1, 63a, CG9969, DOR28, Dmel\CG9969, Dmel_CG9969, Or63aA
Chromosome Location
Chr.3L: 2961534 - 2963788 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035382
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001169860.1 NP_001163331.1 4 - 5 958 76
NM_079171.4 NP_523895.2 5 - 6 885 76

View Full Product Details