Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BtbVII
Entrez Gene ID
38376
Gene Name
BTB-protein-VII
Gene Aliases
BTB-VII, BTBVII, BcDNA:GH07729, CG11494, CG14956, CG43365, Dmel\CG43365, Dmel_CG43365
Chromosome Location
Chr.3L: 3148646 - 3158408 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0263108
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001144408.3 NP_001137880.2 2 - 3 564 76
NM_001259661.2 NP_001246590.1 2 - 3 564 76
NM_001259662.1 NP_001246591.1 2 - 3 542 76
NM_001274426.1 NP_001261355.1 2 - 3 564 76
NM_001274427.2 NP_001261356.1 2 - 3 564 76
NM_079172.4 NP_523896.2 2 - 3 440 76
NM_139517.3 NP_647774.2 2 - 3 564 76
NM_167990.3 NP_728814.1 2 - 3 550 76
GenBank mRNA AY051895.1 - 1 - 2 535 76

View Full Product Details