Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Awh
Entrez Gene ID
38451
Gene Name
Arrowhead
Gene Aliases
CG1072, Dmel\CG1072, Dmel_CG1072, awh, l(3)63Ea, l(3)63Ea/Awh, l(3)64Ea, l(3)SH9, l63Ea
Chromosome Location
Chr.3L: 3846206 - 3861692 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0013751
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274450.1 NP_001261379.1 2 - 3 726 69
NM_001274451.1 NP_001261380.1 2 - 3 726 69
NM_079183.3 NP_523907.2 2 - 3 726 69
NM_168042.2 NP_728906.1 2 - 3 726 69
GenBank mRNA BT025230.1 - 2 - 3 721 69

View Full Product Details