Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
mas
Entrez Gene ID
38499
Gene Name
masquerade
Gene Aliases
CG15002, Dmel\CG15002, Dmel_CG15002, Mas, SPH79, c-SPH79, cSPH79
Chromosome Location
Chr.3L: 4161666 - 4167464 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0011653
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001274479.1 NP_001261408.1 2 - 3 302 70
NM_079195.3 NP_523919.1 2 - 3 302 70
GenBank mRNA BT001597.1 - 2 - 3 299 70

View Full Product Details