Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Sse
Entrez Gene ID
38640
Gene Name
Separase
Gene Aliases
CG10583, Dmel\CG10583, Dmel_CG10583, SSE, l(3)13m-281, l(3)SG14, sse
Chromosome Location
Chr.3L: 5659700 - 5662247 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0035627
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079211.3 NP_523935.1 2 - 3 367 97
GenBank mRNA AY119579.1 - 2 - 3 305 97

View Full Product Details