Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- msl-3
- Entrez Gene ID
-
38779
- Gene Name
- male-specific lethal 3
- Gene Aliases
-
CG8631, DmMsl-3, Dmel\CG8631, Dmel_CG8631, MSL, MSL-3, MSL3, MSL3_DROME, Msl 1-3, Msl3, mle(3)132, mle3, mls3, msl, msl3
- Chromosome Location
-
Chr.3L: 7123213 - 7125533 on Build Release_6_plus_ISO1_MT
- Species Specific ID (Flybase ID)
- FBgn0002775
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_079227.3 |
NP_523951.1 |
4 - 5 |
1453 |
61 |
GenBank mRNA |
AY071074.1 |
- |
4 - 5 |
1443 |
61 |
View Full Product Details