Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
aay
Entrez Gene ID
39085
Gene Name
astray
Gene Aliases
0423/14, 24661601, CG3705, Dmel\CG3705, Dmel_CG3705, anon-WO0172774.117
Chromosome Location
Chr.3L: 9423253 - 9424781 on Build Release_6_plus_ISO1_MT
Species Specific ID (Flybase ID)
FBgn0023129
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_079277.3 NP_524001.2 1 - 1 1441 124
GenBank mRNA AY051689.1 - 1 - 1 1418 124

View Full Product Details